Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01369005_m1
          See other VAC14-AS1 GE Assays ›
          Gene Symbol:
          VAC14-AS1
          Gene Name:
          VAC14 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 70755060 - 70773251 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01369005_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: VAC14-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_034083.2 2 - 3 272 65
          GenBank mRNA AK095020.1 - 2 - 3 198 65
          BC035152.1 - 2 - 3 237 65
          DB487352.1 - 2 - 3 272 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600008283.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16000583.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3355921 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 9965 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 25853 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3355921 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3355926 - -
          Agilent Human 1B Microarray G4111A A_32_P724894 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1564158_PM_a_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR16008533.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16008533.hg.1 NR_034083.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16008533.hg.1 TC16000583.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR16008536.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16008536.hg.1 NR_034083.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16008536.hg.1 TC16000583.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1600010776.hg.1 ENST00000623155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16008535.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16008535.hg.1 NR_034083.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16008535.hg.1 TC16000583.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR16008534.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16008534.hg.1 NR_034083.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16008534.hg.1 TC16000583.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600162484.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162484.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162484.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162484.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600162484.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162484.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162484.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162484.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600162484.hg.1 TC1600008283.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600162486.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162486.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162486.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162486.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600162486.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162486.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162486.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162486.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600162486.hg.1 TC1600008283.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600162488.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162488.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162488.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162488.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600162488.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162488.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162488.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162488.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600162488.hg.1 TC1600008283.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay TC1600008283.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010776.hg.1 ENST00000623155.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162482.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162482.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162482.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162482.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600162482.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162482.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162482.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162482.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600162482.hg.1 TC1600008283.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600162489.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162489.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162489.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162489.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600162489.hg.1 ENST00000398177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162489.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162489.hg.1 ENST00000562507.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600162489.hg.1 NR_034083.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600162489.hg.1 TC1600008283.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1564158_PM_a_at - -
          Human Genome U133 Plus 2.0 Array 1564158_a_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564158_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564158_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564158_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564158_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564158_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564158_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564158_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564158_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564158_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564158_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564158_a_at8 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 228742 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3272838 AK095020.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3272838 BC035152.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3272838 NR_034083.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline