Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01371133_m1
          See other ZNF790-AS1 GE Assays ›
          Gene Symbol:
          ZNF790-AS1
          Gene Name:
          ZNF790 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 36797550 - 36828111 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01371133_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZNF790-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040027.1 3 - 4 359 76
          NR_040028.1 2 - 3 308 76
          GenBank mRNA BM476885.1 - 3 - 4 359 76
          BX394102.2 - 2 - 3 307 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900007953.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000507.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17135235 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3646051 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 16685 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3646051 - -
          Agilent Whole Chr array_18 19 21 A_17_P31579575 - -
          Agilent Whole Chr array_18 19 21 A_17_P10951652 - -
          Agilent Whole Chr array_18 19 21 A_17_P17135216 - -
          Agilent Whole Chr array_18 19 21 A_17_P10951651 - -
          Agilent Whole Chr array_18 19 21 A_17_P31579579 - -
          Agilent Whole Chr array_18 19 21 A_17_P31579580 - -
          Agilent Whole Chr array_18 19 21 A_17_P17135230 - -
          Agilent Whole Chr array_18 19 21 A_17_P17135218 - -
          Agilent Whole Chr array_18 19 21 A_17_P17135235 - -
          Agilent Whole Chr array_18 19 21 A_17_P17135224 - -
          Agilent Whole Chr array_18 19 21 A_17_P17135223 - -
          Agilent Whole Chr array_18 19 21 A_17_P31579553 - -
          Agilent Whole Chr array_18 19 21 A_17_P10951653 - -
          Agilent Whole Chr array_18 19 21 A_17_P17135206 - -
          Agilent Whole Chr array_18 19 21 A_17_P10951654 - -
          Agilent Whole Chr array_18 19 21 A_17_P17135218 - -
          Agilent Whole Chr array_18 19 21 A_17_P17135206 - -
          Agilent Whole Chr array_18 19 21 A_17_P31579579 - -
          Agilent Whole Chr array_18 19 21 A_17_P10951652 - -
          Agilent Whole Chr array_18 19 21 A_17_P17135224 - -
          Agilent Whole Chr array_18 19 21 A_17_P17135216 - -
          Agilent Whole Chr array_18 19 21 A_17_P17135230 - -
          Agilent Whole Chr array_18 19 21 A_17_P31579580 - -
          Agilent Whole Chr array_18 19 21 A_17_P10951651 - -
          Agilent Whole Chr array_18 19 21 A_17_P17135235 - -
          Agilent Whole Chr array_18 19 21 A_17_P10951654 - -
          Agilent Whole Chr array_18 19 21 A_17_P31579575 - -
          Agilent Whole Chr array_18 19 21 A_17_P17135223 - -
          Agilent Whole Chr array_18 19 21 A_17_P10951653 - -
          Agilent Whole Chr array_18 19 21 A_17_P31579553 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 235779_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1558809_PM_s_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR19006709.hg.1 ENST00000500219.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19006709.hg.1 ENST00000501367.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19006709.hg.1 NR_040027.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19006709.hg.1 NR_040028.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19006709.hg.1 TC19000507.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000587278.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000587278.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000587278.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000587278.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000589556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000589556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000589556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 ENST00000589556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 NR_040027.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 NR_040027.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 NR_040027.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 NR_040027.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 NR_040027.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 NR_040027.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 NR_040028.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 NR_040028.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 NR_040028.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007953.hg.1 NR_040028.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19006707.hg.1 ENST00000500219.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19006707.hg.1 ENST00000501367.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19006707.hg.1 NR_040027.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19006707.hg.1 NR_040028.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19006707.hg.1 TC19000507.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19006712.hg.1 ENST00000501367.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19006712.hg.1 NR_040027.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19006712.hg.1 NR_040028.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19006712.hg.1 TC19000507.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19006708.hg.1 NR_040027.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19006708.hg.1 TC19000507.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 ENST00000587278.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 ENST00000589556.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 NR_040027.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 NR_040028.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 TC1900007953.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 ENST00000587278.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 ENST00000589556.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 NR_040027.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900159267.hg.1 NR_040028.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000587278.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000587278.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000587278.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000587278.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000589556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000589556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000589556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 ENST00000589556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 NR_040027.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 NR_040027.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 NR_040027.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 NR_040027.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 NR_040027.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 NR_040027.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 NR_040028.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 NR_040028.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 NR_040028.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007953.hg.1 NR_040028.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900159264.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159264.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159264.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159264.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159264.hg.1 ENST00000589556.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159264.hg.1 NR_040027.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900159264.hg.1 NR_040028.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900159264.hg.1 TC1900007953.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900159264.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159264.hg.1 ENST00000587413.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159264.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159264.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159264.hg.1 ENST00000589556.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159264.hg.1 NR_040027.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900159264.hg.1 NR_040028.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900159266.hg.1 ENST00000587278.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159266.hg.1 ENST00000589556.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159266.hg.1 NR_040027.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900159266.hg.1 TC1900007953.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900159266.hg.1 ENST00000587278.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159266.hg.1 ENST00000589556.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159266.hg.1 NR_040027.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900159274.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159274.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159274.hg.1 NR_040027.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900159274.hg.1 NR_040028.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900159274.hg.1 TC1900007953.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900159274.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159274.hg.1 ENST00000588906.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900159274.hg.1 NR_040027.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900159274.hg.1 NR_040028.1 Refseq
          Human Genome U133 Plus 2.0 Array 235779_PM_at - -
          Human Genome U133 Plus 2.0 Array 1558809_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 235779_at - -
          Human Genome U133 Plus 2.0 Array 1558809_s_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558809_s_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235779_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558809_s_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235779_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235779_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235779_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235779_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558809_s_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558809_s_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558809_s_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235779_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235779_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558809_s_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235779_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558809_s_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558809_s_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558809_s_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235779_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235779_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235779_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558809_s_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558809_s_at7 - -
          Human Genome U95 Set 58060_PM_at - -
          Human Genome U95 Set 58060_at - -
          Human Genome U133 Set 235779_at - -
          Human Genome U133 Set 235779_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A012606 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A012606 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline