| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009089.hg.1 |
ENST00000397549.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009089.hg.1 |
ENST00000397549.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009089.hg.1 |
ENST00000397549.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009089.hg.1 |
ENST00000592718.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009089.hg.1 |
ENST00000592718.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009089.hg.1 |
ENST00000592718.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009089.hg.1 |
NR_109783.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009089.hg.1 |
NR_109783.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009089.hg.1 |
NR_109783.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006824.hg.1 |
ENST00000577230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168646.hg.1 |
ENST00000397549.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168646.hg.1 |
ENST00000592718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168646.hg.1 |
NR_109783.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168646.hg.1 |
ENST00000397549.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168646.hg.1 |
ENST00000592718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168646.hg.1 |
NR_109783.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168646.hg.1 |
TC1700009089.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700168648.hg.1 |
ENST00000397549.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168648.hg.1 |
ENST00000592718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168648.hg.1 |
NR_109783.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168648.hg.1 |
ENST00000397549.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168648.hg.1 |
ENST00000592718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168648.hg.1 |
NR_109783.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168648.hg.1 |
TC1700009089.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00006824.hg.1 |
ENST00000577230.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009089.hg.1 |
ENST00000397549.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009089.hg.1 |
ENST00000397549.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009089.hg.1 |
ENST00000397549.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009089.hg.1 |
ENST00000592718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009089.hg.1 |
ENST00000592718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009089.hg.1 |
ENST00000592718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009089.hg.1 |
NR_109783.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009089.hg.1 |
NR_109783.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009089.hg.1 |
NR_109783.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168647.hg.1 |
ENST00000397549.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168647.hg.1 |
ENST00000592718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168647.hg.1 |
NR_109783.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168647.hg.1 |
ENST00000397549.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168647.hg.1 |
ENST00000592718.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700168647.hg.1 |
NR_109783.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700168647.hg.1 |
TC1700009089.hg.1 |
Clariom D TC |