| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00165306 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00165264 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00165301 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00165315 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00165262 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20189929 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00165314 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20189971 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20189996 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00165265 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00165311 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15053826 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20189989 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00165263 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20189995 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20189930 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20189933 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00165307 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15053829 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15053825 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20189993 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00165302 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20189926 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20189932 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00165300 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P20189928 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00165308 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00165304 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
70711 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
173699 |
NM_002458.1 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
173699 |
NM_017511.1 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
173699 |
S80993.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
173699 |
U78551.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
79295 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
204249 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
703787 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
233083 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
401631 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
611767 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
895065 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
428652 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
46206 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
289511 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
432284 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3680462 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
8253 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P35105958 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P15099943 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P35105958 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_33_P3680462 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_16_P35105958 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P15099943 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P00054226 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P35105958 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
17815 |
- |
- |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
40656 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P15099945 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00054230 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P10102149 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P17542783 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P35105958 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P15099942 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00054226 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P17541716 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P10097859 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P15099943 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00054234 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P35105960 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P00054230 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_18_P10099753 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P00054226 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P35105958 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P15099943 |
- |
- |