| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019671.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019671.hg.1 |
ENST00000543096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019671.hg.1 |
ENST00000543111.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019671.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019671.hg.1 |
TC12001497.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019676.hg.1 |
ENST00000543111.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019676.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019676.hg.1 |
TC12001497.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000539751.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000539751.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000539751.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000543096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000543096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000543096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000543096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000543096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019673.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019673.hg.1 |
ENST00000543111.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019673.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019673.hg.1 |
TC12001497.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019675.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019675.hg.1 |
ENST00000543111.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019675.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019675.hg.1 |
TC12001497.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019670.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019670.hg.1 |
ENST00000543096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019670.hg.1 |
ENST00000543111.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019670.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019670.hg.1 |
TC12001497.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019672.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019672.hg.1 |
ENST00000543111.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019672.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019672.hg.1 |
TC12001497.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019674.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019674.hg.1 |
ENST00000543111.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019674.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12019674.hg.1 |
TC12001497.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200181317.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181317.hg.1 |
ENST00000539751.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181317.hg.1 |
ENST00000543096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181317.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181317.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200181317.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181317.hg.1 |
ENST00000539751.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181317.hg.1 |
ENST00000543096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181317.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181317.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200181317.hg.1 |
TC1200010667.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200181323.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181323.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200181323.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181323.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200181323.hg.1 |
TC1200010667.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000539751.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000539751.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000539751.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000543096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000543096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000543096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000543096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000543096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010667.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200181318.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181318.hg.1 |
ENST00000543096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181318.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181318.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200181318.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181318.hg.1 |
ENST00000543096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181318.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181318.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200181318.hg.1 |
TC1200010667.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200181319.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181319.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181319.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200181319.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181319.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181319.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200181319.hg.1 |
TC1200010667.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200181320.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181320.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181320.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200181320.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181320.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181320.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200181320.hg.1 |
TC1200010667.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200181322.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181322.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181322.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200181322.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181322.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181322.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200181322.hg.1 |
TC1200010667.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200181321.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181321.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181321.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200181321.hg.1 |
ENST00000327337.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181321.hg.1 |
ENST00000543111.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200181321.hg.1 |
NM_001145475.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200181321.hg.1 |
TC1200010667.hg.1 |
Clariom D TC |