Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01373560_g1
          See other LINC00308 GE Assays ›
          Gene Symbol:
          LINC00308
          Gene Name:
          long intergenic non-protein coding RNA 308
          Gene Aliases:
          C21orf74, NCRNA00308, PRED16
          Chromosome Location:
          Chr.21: 22098617 - 22116528 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01373560_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00308
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038400.1 4 - 5 393 87
          GenBank mRNA AY077696.1 - 4 - 5 392 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100006728.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000063.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965824 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11307600 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965896 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965814 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965834 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965829 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965815 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11307568 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965892 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965796 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965866 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11307559 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965845 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965862 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965853 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965844 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965795 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965826 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965800 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965835 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965825 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965868 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965808 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11307570 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965810 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11307584 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965817 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11307612 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11307596 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11307618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965854 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11307586 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965799 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965857 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965876 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965809 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965848 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11307549 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965801 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965869 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965885 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965833 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965832 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965836 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965880 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965839 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11307621 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965891 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965812 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965813 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965840 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965886 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965859 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965889 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965802 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965851 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965881 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965850 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11307530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11307620 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11307550 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31965867 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 22323 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 46430 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 644854 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3307742 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P761454 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 38685 - -
          Agilent Human Genome CGH Microarray 44B A_14_P130897 - -
          Agilent Human Genome CGH Microarray 44A A_14_P130897 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 12583 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3307742 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3307742 - -
          SurePrint G3 Human GE Microarray G4851A A_19_P00800172 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P761454 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P130897 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P130897 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21211605 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P761454 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 16370 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 17442 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P13881329 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21211604 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21211617 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P130897 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21211605 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21211612 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21211599 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21211620 - -
          Agilent Human 1B Microarray G4111A A_32_P761454 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P130897 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8068012 - -
          Affymetrix Human Gene 1.0 ST Array 8068008 - -
          Affymetrix Human Gene 1.0 ST Array 8068007 - -
          Affymetrix Human Gene 1.0 ST Array 8068011 - -
          Affymetrix Human Gene 1.0 ST Array 8068009 - -
          Affymetrix Human Gene 1.0 ST Array 8068013 - -
          Affymetrix Human Gene 1.0 ST Array 8068010 - -
          Affymetrix Human Gene 1.1 ST Array 8068006 - -
          Affymetrix HT HG-U133+ PM Array Plate 1552605_PM_s_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1552604_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2100006728.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006728.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006728.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006728.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006728.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006728.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006728.hg.1 NR_038400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006728.hg.1 NR_038400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006728.hg.1 NR_038400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006728.hg.1 NR_038400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006728.hg.1 NR_038400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006728.hg.1 NR_038400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000327.hg.1 ENST00000419431.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000327.hg.1 NR_038400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000327.hg.1 TC21000063.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21000324.hg.1 ENST00000419431.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000324.hg.1 NR_038400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000324.hg.1 TC21000063.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21000320.hg.1 ENST00000419431.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000320.hg.1 NR_038400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000320.hg.1 TC21000063.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21000323.hg.1 ENST00000419431.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000323.hg.1 NR_038400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000323.hg.1 TC21000063.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21000325.hg.1 ENST00000419431.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000325.hg.1 NR_038400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000325.hg.1 TC21000063.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21000326.hg.1 ENST00000419431.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000326.hg.1 NR_038400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000326.hg.1 TC21000063.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2100145493.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145493.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145493.hg.1 TC2100006728.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100145493.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145493.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145496.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145496.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145496.hg.1 TC2100006728.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100145496.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145496.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145489.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145489.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145489.hg.1 TC2100006728.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100145489.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145489.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145492.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145492.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145492.hg.1 TC2100006728.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100145492.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145492.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145495.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145495.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145495.hg.1 TC2100006728.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100145495.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145495.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145494.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145494.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145494.hg.1 TC2100006728.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100145494.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145494.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006728.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006728.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006728.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006728.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006728.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006728.hg.1 ENST00000419431.2 Ensembl
          Affymetrix Human Clariom D Assay TC2100006728.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006728.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006728.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006728.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006728.hg.1 NR_038400.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006728.hg.1 NR_038400.1 Refseq
          Human Genome U133 Plus 2.0 Array 1552604_at - -
          Human Genome U133 Plus 2.0 Array 1552604_PM_at - -
          Human Genome U133 Plus 2.0 Array 1552605_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 1552605_s_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552605_s_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552605_s_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552604_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552604_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552605_s_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552604_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552604_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552605_s_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552605_s_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552604_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552604_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552604_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552604_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552605_s_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552605_s_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552604_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552604_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552604_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552605_s_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552605_s_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552605_s_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1552605_s_at6 - -
          Human Genome U95 Set 55120_at - -
          Human Genome U95 Set 55120_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019096 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019096 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 228320 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1716149 NR_038400.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1716149 AY077696.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1716149 NR_038400.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1716149 AY077696.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1716149 NR_038400.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1716149 AY077696.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8fhbl:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0