| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019681.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019681.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019681.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019681.hg.1 |
TC11001575.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000527960.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000527960.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000527960.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000527960.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000534287.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000534287.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000534287.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000534287.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
ENST00000534287.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010616.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019679.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019679.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11019679.hg.1 |
TC11001575.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100180398.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180398.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180398.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180398.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180398.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180398.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180398.hg.1 |
TC1100010616.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100180400.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180400.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180400.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180400.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180400.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180400.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180400.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180400.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180400.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180400.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180400.hg.1 |
TC1100010616.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000499194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000527960.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000527960.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000527960.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000527960.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000528285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000534287.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000534287.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000534287.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000534287.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
ENST00000534287.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
NR_038907.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010616.hg.1 |
NR_038907.1 |
Refseq |
| Human Genome U133 Plus 2.0 Array |
1557565_PM_a_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1557564_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1557565_a_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1557564_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557564_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557565_a_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557564_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557564_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557565_a_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557565_a_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557565_a_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557565_a_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557564_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557565_a_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557564_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557564_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557565_a_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557564_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557564_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557565_a_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557564_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557564_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557565_a_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557564_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557565_a_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1557565_a_at3 |
- |
- |