| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
1569322_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10006273.hg.1 |
ENST00000422847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10006273.hg.1 |
NR_038464.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10006273.hg.1 |
TC10000577.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10006278.hg.1 |
ENST00000422847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10006278.hg.1 |
NR_038464.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10006278.hg.1 |
TC10000577.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008226.hg.1 |
ENST00000422847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008226.hg.1 |
ENST00000422847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008226.hg.1 |
ENST00000422847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008226.hg.1 |
ENST00000432308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008226.hg.1 |
NR_038464.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008226.hg.1 |
NR_038464.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008226.hg.1 |
NR_038464.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10006277.hg.1 |
ENST00000422847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10006277.hg.1 |
NR_038464.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10006277.hg.1 |
TC10000577.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000155711.hg.1 |
ENST00000422847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155711.hg.1 |
ENST00000432308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155711.hg.1 |
NR_038464.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000155711.hg.1 |
ENST00000422847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155711.hg.1 |
ENST00000432308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155711.hg.1 |
NR_038464.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000155711.hg.1 |
TC1000008226.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000155712.hg.1 |
ENST00000422847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155712.hg.1 |
NR_038464.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000155712.hg.1 |
ENST00000422847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155712.hg.1 |
NR_038464.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000155712.hg.1 |
TC1000008226.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000155708.hg.1 |
ENST00000422847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155708.hg.1 |
NR_038464.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000155708.hg.1 |
ENST00000422847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000155708.hg.1 |
NR_038464.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000155708.hg.1 |
TC1000008226.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000008226.hg.1 |
ENST00000422847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008226.hg.1 |
ENST00000422847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008226.hg.1 |
ENST00000422847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008226.hg.1 |
ENST00000432308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008226.hg.1 |
NR_038464.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008226.hg.1 |
NR_038464.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008226.hg.1 |
NR_038464.1 |
Refseq |
| Human Genome U133 Plus 2.0 Array |
1569322_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1569322_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1569322_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1569322_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1569322_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1569322_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1569322_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1569322_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1569322_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1569322_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1569322_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1569322_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1569322_at9 |
- |
- |