| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013158.hg.1 |
ENST00000628310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010459.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010459.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010459.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010459.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010459.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010459.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010459.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010459.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010459.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010459.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174416.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174416.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174416.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174416.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174416.hg.1 |
TC0700010459.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700174418.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174418.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174418.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174418.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174418.hg.1 |
TC0700010459.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300013158.hg.1 |
ENST00000628310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174417.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174417.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174417.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174417.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174417.hg.1 |
TC0700010459.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700174419.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174419.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174419.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174419.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174419.hg.1 |
TC0700010459.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700010459.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010459.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010459.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010459.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010459.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010459.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010459.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010459.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010459.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010459.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174415.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174415.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174415.hg.1 |
ENST00000419813.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700174415.hg.1 |
NR_046220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700174415.hg.1 |
TC0700010459.hg.1 |
Clariom D TC |