Search
Search

LOC105378614| Gene Symbol: LOC105378614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135054.1 | 1 - 2 | 123 | 71 | ||
| GenBank mRNA | AI791463.1 | - | 1 - 2 | 153 | 71 | |
| BX106629.1 | - | 1 - 2 | 142 | 71 | ||
Set Membership: |
|