| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
ENST00000462263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
ENST00000462263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
ENST00000462263.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010396.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07018370.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07018370.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07018370.hg.1 |
TC07001177.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07018371.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07018371.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07018371.hg.1 |
TC07001177.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700173911.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700173911.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700173911.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700173911.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700173911.hg.1 |
TC0700010396.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700173910.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700173910.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700173910.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700173910.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700173910.hg.1 |
TC0700010396.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
ENST00000222567.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
ENST00000462263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
ENST00000462263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
ENST00000462263.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
NM_001002926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010396.hg.1 |
NM_001002926.1 |
Refseq |