Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01380312_s1
          See other KRT8P41 GE Assays ›
          Gene Symbol:
          KRT8P41
          Gene Name:
          keratin 8 pseudogene 41
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 9094363 - 9096190 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01380312_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KRT8P41
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027713.1 1 1370 125
          GenBank mRNA AK127993.1 - 1 1370 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100006802.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000168.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07631006 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 376369 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3605269 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P489690 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 1683 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 8988 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 15440 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3605269 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3605269 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07631000 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16418596 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07631001 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07630997 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28030066 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07630999 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28030046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16418594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28030069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16418612 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28030048 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16418608 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07630996 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16418598 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28030071 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16418617 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28030048 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28030071 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16418598 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07630997 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16418608 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16418594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28030066 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28030069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07631001 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16418612 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07630996 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16418617 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16418596 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28030046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07630999 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07631000 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P489690 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P489695 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P489690 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 2885 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07630975 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07630989 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07630984 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16418594 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16418582 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16418586 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07631006 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07631003 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07630978 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07630979 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16418580 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16418600 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16418585 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07630987 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07630994 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16418596 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16418598 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07630960 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07630995 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07630991 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07630966 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07631000 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16418591 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16418588 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16418584 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07631004 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07630962 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07631005 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07630983 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07630996 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07630971 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7938312 - -
          Affymetrix Human Gene 1.0 ST Array 7938310 - -
          Affymetrix Human Gene 1.0 ST Array 7938311 - -
          Affymetrix Human Gene 1.1 ST Array 7938309 - -
          Affymetrix Human Transcriptome Array 2.0 TC1100006802.hg.1 ENST00000533985.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006802.hg.1 NR_027713.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006802.hg.1 NR_027713.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11002302.hg.1 NR_027713.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11002302.hg.1 TC11000168.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100147713.hg.1 ENST00000533985.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100147713.hg.1 NR_027713.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100147713.hg.1 ENST00000533985.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100147713.hg.1 NR_027713.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100147713.hg.1 TC1100006802.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100006802.hg.1 ENST00000533985.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006802.hg.1 NR_027713.1 Refseq
          Affymetrix Human Clariom D Assay TC1100006802.hg.1 NR_027713.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100147714.hg.1 NR_027713.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100147714.hg.1 NR_027713.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100147714.hg.1 TC1100006802.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046122 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046121 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046121 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046122 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 229058 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline