Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01381837_m1
          See other LINC00551 GE Assays ›
          Gene Symbol:
          LINC00551
          Gene Name:
          long intergenic non-protein coding RNA 551
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 106617810 - 106631658 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01381837_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00551
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_051977.1 3 - 4 1097 70
          GenBank mRNA AK098538.1 - 3 - 4 1096 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300009995.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562626 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113683 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562676 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562582 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562642 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562560 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562629 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562554 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113651 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113640 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562605 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562658 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562701 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562696 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113659 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113699 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562663 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113649 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562695 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562702 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16667775 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562609 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562601 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562553 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113675 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562644 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113644 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113599 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562578 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113713 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562660 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113585 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113645 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562693 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113666 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562641 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562632 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562669 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562599 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113691 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562699 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113688 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16667771 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113714 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113660 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562697 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16667772 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16667767 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113575 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113684 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562608 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562700 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562627 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562672 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562564 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562674 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113663 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562550 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113658 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113667 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113630 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562659 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113619 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113662 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562586 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562661 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113602 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562619 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562649 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562704 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113593 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113605 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113686 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562675 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562690 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113652 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562677 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113617 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113607 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113621 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113676 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562607 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562703 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562585 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113589 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113708 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113631 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562698 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16667778 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562543 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562628 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562686 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113718 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562551 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16667776 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113669 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113736 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562651 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113709 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113712 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562621 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562692 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29562647 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113600 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09113696 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 99744 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 50431 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 700057 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 526713 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 765138 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 310940 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 146952 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3679936 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 46623 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P19948346 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P19948346 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3679936 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3679936 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P19948346 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P19948346 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 165835 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 57328 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19948336 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19948346 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P40088425 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02858432 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P19948346 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 244119_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1300009995.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009995.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009995.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009995.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009995.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009995.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009995.hg.1 NR_051977.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009995.hg.1 NR_051977.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009995.hg.1 NR_051977.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009995.hg.1 NR_051977.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009995.hg.1 NR_051977.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009995.hg.1 NR_051977.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009995.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167031.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167031.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167031.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167031.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167031.hg.1 TC1300009995.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300167038.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167038.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167038.hg.1 TC1300009995.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300167032.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167032.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167032.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167032.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167032.hg.1 TC1300009995.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300167036.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167036.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167036.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167036.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167036.hg.1 TC1300009995.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1300009995.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009995.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009995.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009995.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009995.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009995.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009995.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009995.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009995.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009995.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009995.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009995.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009995.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167033.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167033.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167033.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167033.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167033.hg.1 TC1300009995.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300167030.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167030.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167030.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167030.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167030.hg.1 TC1300009995.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300167037.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167037.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167037.hg.1 ENST00000607593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167037.hg.1 NR_051977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300167037.hg.1 TC1300009995.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 244119_at - -
          Human Genome U133 Plus 2.0 Array 244119_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244119_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244119_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244119_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244119_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244119_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244119_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244119_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244119_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244119_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244119_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244119_at6 - -
          Human Genome U95 Set 70208_PM_at - -
          Human Genome U95 Set 70208_at - -
          Human Genome U133 Set 244119_at - -
          Human Genome U133 Set 244119_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 233120 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline