| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012776.hg.1 |
ENST00000545185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012776.hg.1 |
ENST00000545185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012776.hg.1 |
ENST00000545185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012776.hg.1 |
ENST00000545185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14006475.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14006475.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14006475.hg.1 |
TC14000517.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007872.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007872.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007872.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007872.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007872.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007872.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007872.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007872.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007872.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007872.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14006474.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14006474.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14006474.hg.1 |
TC14000517.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1400155761.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400155761.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400155761.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400155761.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400155761.hg.1 |
TC1400007872.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400155762.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400155762.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400155762.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400155762.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400155762.hg.1 |
TC1400007872.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1400007872.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007872.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007872.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007872.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007872.hg.1 |
ENST00000557527.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007872.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007872.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007872.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007872.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007872.hg.1 |
NR_038445.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012776.hg.1 |
ENST00000545185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012776.hg.1 |
ENST00000545185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012776.hg.1 |
ENST00000545185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012776.hg.1 |
ENST00000545185.1 |
Ensembl |