Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01382362_g1
          A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



          Species:
          Human
          Assay Type:
          Non-coding
          Assay ID Hs01382362_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details


          Back To Top

          More Information


          Set Membership:

          Homo sapiens 1700

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23034068 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02888620 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02888610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02888625 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23034002 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02888632 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23034067 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02888623 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02888618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23034054 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02888629 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02888609 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15532319 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15532298 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23034049 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23034055 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15532323 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23034063 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23034070 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02888614 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02888631 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02888613 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3217764 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 129208 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 72066 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P23419293 - -
          Agilent Human 1B Microarray G4111A A_32_P939217 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8093455 - -
          Affymetrix Human Gene 1.0 ST Array 8093454 - -
          Affymetrix Human Gene 1.1 ST Array 8093453 - -
          Affymetrix HT HG-U133+ PM Array Plate 1564152_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0400006504.hg.1 ENST00000417557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400006504.hg.1 ENST00000417557.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006504.hg.1 ENST00000417557.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400006504.hg.1 ENST00000417557.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1564152_at - -
          Human Genome U133 Plus 2.0 Array 1564152_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564152_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564152_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564152_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564152_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564152_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564152_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564152_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564152_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564152_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564152_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564152_at11 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 703319 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1805011 BC121811.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1805011 AK093135.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1783082 - -
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1805011 BC121811.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1805011 AK093135.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1783082 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1783082 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1805011 BC121811.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1805011 AK093135.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1805011 BC121811.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1805011 AK093135.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1805011 BC121811.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1805011 AK093135.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1805011 BC121811.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1805011 AK093135.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline