| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000529171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000529171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000529171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000529171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000529171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000529171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000532515.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000532515.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000532515.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000532515.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000553269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000553269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000553269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000553269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000553269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000553269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000553775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009342.hg.1 |
ENST00000553775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000529171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000529171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000529171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000529171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000529171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000529171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000532515.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000532515.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000532515.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000532515.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000553269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000553269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000553269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000553269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000553269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000553269.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000553775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009342.hg.1 |
ENST00000553775.1 |
Ensembl |