| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000425157.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000425157.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000425157.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000425157.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000455051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000455051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000455051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
ENST00000455051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900175348.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900175348.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900175348.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900175348.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900175348.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900175348.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900175348.hg.1 |
TC0900011050.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000425157.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000425157.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000425157.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000425157.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000455051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000455051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000455051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
ENST00000455051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011050.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900175345.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900175345.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900175345.hg.1 |
ENST00000455051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900175345.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900175345.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900175345.hg.1 |
ENST00000411575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900175345.hg.1 |
ENST00000455051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900175345.hg.1 |
NR_121578.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900175345.hg.1 |
TC0900011050.hg.1 |
Clariom D TC |