| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8030839 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8038875 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8038876 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8038874 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8030838 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1559093_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024233.hg.1 |
ENST00000301439.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024233.hg.1 |
NR_034159.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024233.hg.1 |
TC19001783.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024232.hg.1 |
ENST00000301439.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024232.hg.1 |
NR_034159.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024232.hg.1 |
TC19001783.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900194842.hg.1 |
NR_034159.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194842.hg.1 |
NR_034159.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194842.hg.1 |
TC1900011265.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900194843.hg.1 |
NR_034159.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194843.hg.1 |
NR_034159.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194843.hg.1 |
TC1900011265.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900194841.hg.1 |
NR_034159.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194841.hg.1 |
NR_034159.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194841.hg.1 |
TC1900011265.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900194840.hg.1 |
NR_034159.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194840.hg.1 |
NR_034159.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194840.hg.1 |
TC1900011265.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
1559093_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1559093_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559093_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559093_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559093_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559093_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559093_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559093_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559093_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559093_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559093_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559093_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1559093_at6 |
- |
- |