Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01387417_m1
          See other LINC00485 GE Assays ›
          Gene Symbol:
          LINC00485
          Gene Name:
          long intergenic non-protein coding RNA 485
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 102809283 - 102824399 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01387417_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00485
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033855.1 1 - 2 103 78
          GenBank mRNA BC037211.1 - 1 - 2 103 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200011730.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001891.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580626 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013789 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580572 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013814 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013902 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013809 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013886 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013805 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013863 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580536 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580589 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580506 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580513 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580496 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580497 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013815 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013759 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013848 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013766 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580577 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013813 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16586382 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013859 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580627 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013762 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580576 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580619 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013851 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013752 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580608 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013841 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580633 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580552 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013780 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580607 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013798 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013773 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580512 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580614 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013893 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013794 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580558 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580557 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580489 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013821 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013832 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580518 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580504 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013856 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580521 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013769 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580592 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013807 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580516 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580510 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580505 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013892 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16586384 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013785 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580493 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013801 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580531 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013800 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580503 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580628 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580564 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580515 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580563 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580582 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16586388 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013799 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013824 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580570 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580630 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013771 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580625 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013820 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580603 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580500 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580612 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580621 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013866 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580554 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013808 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580588 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580616 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580520 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580602 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580609 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013888 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013837 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580502 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013890 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16586391 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013817 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013840 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013868 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08580568 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013806 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013822 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013860 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29013796 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 171499 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 41660 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 29743 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 266777 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 541673 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 402272 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02691188 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02691188 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P02691188 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02691188 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 175641 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 47183 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P11583185 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P39796469 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02691193 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19662600 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02691188 - -
          Agilent Human 1B Microarray G4111A A_32_P445535 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P39796469 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02691188 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1561222_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR12024706.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12024706.hg.1 NR_033855.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12024706.hg.1 TC12001891.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12024707.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12024707.hg.1 NR_033855.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12024707.hg.1 TC12001891.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200011730.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011730.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011730.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011730.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011730.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011730.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011730.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011730.hg.1 NR_033855.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011730.hg.1 NR_033855.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011730.hg.1 NR_033855.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011730.hg.1 NR_033855.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011730.hg.1 NR_033855.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011730.hg.1 NR_033855.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011730.hg.1 NR_033855.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190883.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190883.hg.1 NR_033855.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190883.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190883.hg.1 NR_033855.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190883.hg.1 TC1200011730.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200190884.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190884.hg.1 NR_033855.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190884.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190884.hg.1 NR_033855.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190884.hg.1 TC1200011730.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200011730.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011730.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011730.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011730.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011730.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011730.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011730.hg.1 ENST00000547179.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011730.hg.1 NR_033855.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011730.hg.1 NR_033855.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011730.hg.1 NR_033855.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011730.hg.1 NR_033855.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011730.hg.1 NR_033855.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011730.hg.1 NR_033855.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011730.hg.1 NR_033855.1 Refseq
          Human Genome U133 Plus 2.0 Array 1561222_at - -
          Human Genome U133 Plus 2.0 Array 1561222_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561222_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561222_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561222_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561222_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561222_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561222_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561222_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561222_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561222_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561222_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561222_at4 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047950 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047950 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 340758 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1737230 BC037211.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1737230 NR_033855.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1737230 BC037211.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1737230 NR_033855.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1737230 BC037211.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1737230 NR_033855.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1737230 BC037211.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1737230 NR_033855.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1737230 BC037211.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1737230 NR_033855.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline