Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01387463_g1
          See other HES5 GE Assays ›
          Gene Symbol:
          HES5
          Gene Name:
          hes family bHLH transcription factor 5
          Gene Aliases:
          bHLHb38
          Chromosome Location:
          Chr.1: 2528745 - 2530554 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01387463_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HES5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001010926.3 NP_001010926.1 1 - 2 137 62
          XM_005244751.4 XP_005244808.1 1 - 2 446 62
          GenBank mRNA DQ272660.1 - 1 - 2 137 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100012535.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002121.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HES5

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          chondrocyte differentiation
          transcription, DNA-templated
          protein complex assembly
          cell adhesion
          Notch signaling pathway
          smoothened signaling pathway
          brain development
          positive regulation of cell proliferation
          oligodendrocyte development
          telencephalon development
          glial cell fate commitment
          forebrain radial glial cell differentiation
          neural tube development
          central nervous system myelination
          positive regulation of BMP signaling pathway
          regulation of myelination
          auditory receptor cell differentiation
          positive regulation of tyrosine phosphorylation of Stat3 protein
          auditory receptor cell fate determination
          camera-type eye development
          regulation of cell differentiation
          regulation of auditory receptor cell differentiation
          negative regulation of neuron differentiation
          positive regulation of Notch signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JAK-STAT cascade
          cell maturation
          positive regulation of smooth muscle cell proliferation
          astrocyte differentiation
          negative regulation of astrocyte differentiation
          negative regulation of oligodendrocyte differentiation
          regulation of epithelial cell proliferation
          regulation of neurogenesis
          cartilage development
          inner ear receptor stereocilium organization
          comma-shaped body morphogenesis
          S-shaped body morphogenesis
          specification of loop of Henle identity
          metanephric nephron tubule morphogenesis
          establishment of epithelial cell polarity
          neuronal stem cell population maintenance
          negative regulation of stem cell differentiation
          negative regulation of pro-B cell differentiation
          negative regulation of forebrain neuron differentiation
          negative regulation of inner ear receptor cell differentiation
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          chromatin binding
          double-stranded DNA binding
          transcription factor binding
          protein dimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100012535.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012535.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012535.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012535.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012535.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012535.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012535.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012535.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012535.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100012535.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01032602.hg.1 TC01002121.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01032602.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01032602.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01032600.hg.1 TC01002121.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01032600.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01032600.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100192766.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192766.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100192766.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192766.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100192766.hg.1 TC0100012535.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100012535.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100012535.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100012535.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100012535.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100012535.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100012535.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Clariom D Assay TC0100012535.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Clariom D Assay TC0100012535.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Clariom D Assay TC0100012535.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Clariom D Assay TC0100012535.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100192763.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192763.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100192763.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192763.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100192763.hg.1 TC0100012535.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100192765.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192765.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100192765.hg.1 ENST00000378453.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100192765.hg.1 NM_001010926.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100192765.hg.1 TC0100012535.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline