Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01387769_m1
          See other LINC01342 GE Assays ›
          Gene Symbol:
          LINC01342
          Gene Name:
          long intergenic non-protein coding RNA 1342
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 1137017 - 1144056 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01387769_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01342
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038869.1 2 - 3 304 105
          GenBank mRNA BC028014.1 - 2 - 3 304 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100006490.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000026.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20004598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000993 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001027 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001056 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001024 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001003 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15007533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15007508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15007488 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20004614 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000974 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000973 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15007489 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000986 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001006 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000985 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001045 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15007510 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000969 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000982 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20004618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001001 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001038 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20004566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001052 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000971 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15007558 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20004580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001012 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000990 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001008 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001019 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001020 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000972 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20004561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001017 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001058 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001011 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001036 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000984 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15007542 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000994 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001014 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000983 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001023 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000977 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20004610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001049 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15007549 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001007 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001057 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20004506 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15007546 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000967 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000996 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001033 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20004490 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000988 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15007540 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000989 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000997 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001005 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001054 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15007507 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20004602 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001000 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001046 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20004510 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001031 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20004461 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00000979 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001009 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001002 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20004574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20004582 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20004591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001039 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00001044 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 153750 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 176138 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 479576 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 31713 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 372394 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 162067 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 165247 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 24350 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 415042 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 164612 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 280073 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 185868 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 354765 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 341113 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3468017 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 16609 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3468017 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3468017 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P15002253 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 112618 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P35002469 - -
          Agilent Human 1B Microarray G4111A A_32_P341787 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P35002469 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1562788_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR01000302.hg.1 ENST00000416774.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01000302.hg.1 NR_038869.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01000302.hg.1 TC01000026.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100006490.hg.1 ENST00000416774.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006490.hg.1 ENST00000416774.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006490.hg.1 ENST00000416774.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006490.hg.1 NR_038869.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006490.hg.1 NR_038869.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006490.hg.1 NR_038869.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01000301.hg.1 ENST00000416774.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01000301.hg.1 NR_038869.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01000301.hg.1 TC01000026.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01000303.hg.1 ENST00000416774.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01000303.hg.1 NR_038869.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01000303.hg.1 TC01000026.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0100006490.hg.1 ENST00000416774.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006490.hg.1 ENST00000416774.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006490.hg.1 ENST00000416774.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006490.hg.1 NR_038869.1 Refseq
          Affymetrix Human Clariom D Assay TC0100006490.hg.1 NR_038869.1 Refseq
          Affymetrix Human Clariom D Assay TC0100006490.hg.1 NR_038869.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100144602.hg.1 ENST00000416774.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100144602.hg.1 NR_038869.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100144602.hg.1 ENST00000416774.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100144602.hg.1 NR_038869.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100144602.hg.1 TC0100006490.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100144601.hg.1 ENST00000416774.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100144601.hg.1 NR_038869.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100144601.hg.1 ENST00000416774.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100144601.hg.1 NR_038869.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100144601.hg.1 TC0100006490.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100144603.hg.1 ENST00000416774.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100144603.hg.1 NR_038869.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100144603.hg.1 ENST00000416774.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100144603.hg.1 NR_038869.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100144603.hg.1 TC0100006490.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1562788_PM_at - -
          Human Genome U133 Plus 2.0 Array 1562788_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562788_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562788_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562788_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562788_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562788_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562788_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562788_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562788_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562788_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562788_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1562788_at10 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014194 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014194 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 230388 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1823491 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 1850364 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_89084 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1817686 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1817686 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1823491 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-djsrz:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline