Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01388268_m1
          See other FREM2 GE Assays ›
          Gene Symbol:
          FREM2
          Gene Name:
          FRAS1 related extracellular matrix protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 38687036 - 38887131 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01388268_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FREM2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207361.5 NP_997244.4 1 - 2 5486 76
          XM_017020554.1 XP_016876043.1 1 - 2 5481 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300006919.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000132.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FREM2

          Gene Ontology Categories:

          Function(s) Process(es)

          morphogenesis of an epithelium
          cell communication
          cell adhesion
          heart development
          inner ear development
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR13001519.hg.1 ENST00000280481.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13001519.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13001519.hg.1 TC13000132.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000482551.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000482551.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000482551.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 ENST00000482551.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13001518.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13001518.hg.1 TC13000132.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR13001520.hg.1 ENST00000280481.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13001520.hg.1 NM_207361.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13001520.hg.1 TC13000132.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1300147000.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay PSR1300147000.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay PSR1300147000.hg.1 TC1300006919.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300147001.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300147001.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay PSR1300147001.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300147001.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay PSR1300147001.hg.1 TC1300006919.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300147004.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300147004.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay PSR1300147004.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300147004.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay PSR1300147004.hg.1 TC1300006919.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300147003.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300147003.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay PSR1300147003.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300147003.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay PSR1300147003.hg.1 TC1300006919.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000280481.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000482551.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000482551.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000482551.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 ENST00000482551.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq
          Affymetrix Human Clariom D Assay TC1300006919.hg.1 NM_207361.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline