| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000551174.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000551174.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000551174.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000551174.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000551174.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
ENST00000551174.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200163791.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163791.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163791.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200163791.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163791.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163791.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200163791.hg.1 |
TC1200008658.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200163788.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163788.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163788.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200163788.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163788.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163788.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200163788.hg.1 |
TC1200008658.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200163790.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163790.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163790.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200163790.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163790.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163790.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200163790.hg.1 |
TC1200008658.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000551174.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000551174.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000551174.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000551174.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000551174.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
ENST00000551174.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008658.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200163789.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163789.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163789.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200163789.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163789.hg.1 |
ENST00000550185.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200163789.hg.1 |
NR_110103.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200163789.hg.1 |
TC1200008658.hg.1 |
Clariom D TC |