| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011314.hg.1 |
ENST00000356815.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011314.hg.1 |
ENST00000356815.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011314.hg.1 |
ENST00000356815.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011314.hg.1 |
ENST00000356815.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011314.hg.1 |
NM_001003938.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011314.hg.1 |
NM_001003938.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011314.hg.1 |
NM_001003938.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011314.hg.1 |
NM_001003938.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000069.hg.1 |
TC16002033.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000069.hg.1 |
ENST00000356815.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000069.hg.1 |
NM_001003938.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000068.hg.1 |
TC16002033.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000068.hg.1 |
ENST00000356815.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000068.hg.1 |
NM_001003938.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600188417.hg.1 |
ENST00000356815.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600188417.hg.1 |
NM_001003938.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600188417.hg.1 |
ENST00000356815.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600188417.hg.1 |
NM_001003938.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600188417.hg.1 |
TC1600011314.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600188418.hg.1 |
ENST00000356815.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600188418.hg.1 |
NM_001003938.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600188418.hg.1 |
ENST00000356815.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600188418.hg.1 |
NM_001003938.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600188418.hg.1 |
TC1600011314.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1600011314.hg.1 |
ENST00000356815.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011314.hg.1 |
ENST00000356815.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011314.hg.1 |
ENST00000356815.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011314.hg.1 |
ENST00000356815.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011314.hg.1 |
NM_001003938.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011314.hg.1 |
NM_001003938.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011314.hg.1 |
NM_001003938.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011314.hg.1 |
NM_001003938.3 |
Refseq |