| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000540237.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000540237.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000540237.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009688.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200172281.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200172281.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200172281.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200172281.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200172281.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200172281.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200172281.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200172281.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200172281.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200172281.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200172281.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200172281.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200172281.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200172281.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200172281.hg.1 |
TC1200009688.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200172287.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200172287.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200172287.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200172287.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200172287.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200172287.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200172287.hg.1 |
TC1200009688.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000540237.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000540237.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000540237.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000540619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
ENST00000544717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009688.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200172286.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200172286.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200172286.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200172286.hg.1 |
ENST00000536518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200172286.hg.1 |
NR_120476.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200172286.hg.1 |
NR_120477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200172286.hg.1 |
TC1200009688.hg.1 |
Clariom D TC |