Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01394182_m1
          See other TMEM221 GE Assays ›
          Gene Symbol:
          TMEM221
          Gene Name:
          transmembrane protein 221
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 17435509 - 17448839 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01394182_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TMEM221
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001190844.1 NP_001177773.1 2 - 3 510 66
          XM_011527603.2 XP_011525905.1 2 - 3 782 66
          GenBank mRNA BY797043.2 - 2 - 3 510 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900011905.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19001276.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902131 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902124 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902110 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902111 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31520204 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902133 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31520214 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31520207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902122 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902126 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902123 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902117 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902118 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31520219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902128 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31520197 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31520217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902120 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902121 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902132 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31520212 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31520195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10902112 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 61549 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3411612 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20971034 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20971038 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8035314 - -
          Affymetrix Human Gene 1.0 ST Array 8035313 - -
          Affymetrix Human Gene 1.1 ST Array 8035311 - -
          Affymetrix HT HG-U133+ PM Array Plate 1559687_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR19018062.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19018062.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19018062.hg.1 TC19001276.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19018060.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19018060.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19018060.hg.1 TC19001276.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19018063.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19018063.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19018063.hg.1 TC19001276.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 ENST00000593461.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 ENST00000593461.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 ENST00000593461.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011905.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900205158.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205158.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900205158.hg.1 TC1900011905.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900205158.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205158.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900205164.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205164.hg.1 ENST00000593461.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205164.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900205164.hg.1 TC1900011905.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900205164.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205164.hg.1 ENST00000593461.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205164.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900205166.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205166.hg.1 ENST00000593461.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205166.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900205166.hg.1 TC1900011905.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900205166.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205166.hg.1 ENST00000593461.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205166.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 ENST00000593461.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 ENST00000593461.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 ENST00000593461.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011905.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900205159.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205159.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900205159.hg.1 TC1900011905.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900205159.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205159.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900205160.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205160.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900205160.hg.1 TC1900011905.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900205160.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205160.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900205169.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205169.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900205169.hg.1 TC1900011905.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900205169.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205169.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900205170.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205170.hg.1 NM_001190844.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900205170.hg.1 TC1900011905.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900205170.hg.1 ENST00000341130.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900205170.hg.1 NM_001190844.1 Refseq
          Human Genome U133 Plus 2.0 Array 1559687_at - -
          Human Genome U133 Plus 2.0 Array 1559687_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559687_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559687_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559687_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559687_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559687_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559687_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559687_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559687_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559687_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559687_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559687_at9 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 212831 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline