| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
228832_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04007698.hg.1 |
TC04000523.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04007698.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04007698.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04007699.hg.1 |
TC04000523.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04007699.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04007699.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008254.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008254.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008254.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008254.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008254.hg.1 |
ENST00000529296.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008254.hg.1 |
ENST00000529296.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008254.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008254.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008254.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008254.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400156800.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156800.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400156800.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156800.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400156800.hg.1 |
TC0400008254.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400156799.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156799.hg.1 |
ENST00000529296.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156799.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400156799.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156799.hg.1 |
ENST00000529296.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156799.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400156799.hg.1 |
TC0400008254.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400156802.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156802.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400156802.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156802.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400156802.hg.1 |
TC0400008254.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400156801.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156801.hg.1 |
ENST00000529296.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156801.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400156801.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156801.hg.1 |
ENST00000529296.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156801.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400156801.hg.1 |
TC0400008254.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400008254.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008254.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008254.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008254.hg.1 |
ENST00000527564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008254.hg.1 |
ENST00000529296.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008254.hg.1 |
ENST00000529296.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008254.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008254.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008254.hg.1 |
NR_033874.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008254.hg.1 |
NR_033874.1 |
Refseq |
| Human Genome U133 Plus 2.0 Array |
228832_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
228832_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228832_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228832_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228832_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228832_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228832_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228832_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228832_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228832_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228832_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228832_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228832_at7 |
- |
- |
| Human Genome U95 Set |
51049_at |
- |
- |
| Human Genome U95 Set |
51049_PM_at |
- |
- |
| Human Genome U133 Set |
228832_PM_at |
- |
- |
| Human Genome U133 Set |
228832_at |
- |
- |