| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023941.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023941.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023941.hg.1 |
TC11001993.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011347.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100187531.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100187531.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100187531.hg.1 |
ENST00000335385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100187531.hg.1 |
NM_005995.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100187531.hg.1 |
TC1100011347.hg.1 |
Clariom D TC |