Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01547324_gH
          See other PYCARD GE Assays ›
          Gene Symbol:
          PYCARD
          Gene Name:
          PYD and CARD domain containing
          Gene Aliases:
          ASC, CARD5, TMS, TMS-1, TMS1
          Chromosome Location:
          Chr.16: 31201486 - 31202776 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01547324_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PYCARD
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013258.4 NP_037390.2 1 - 2 361 103
          GenBank mRNA AB023416.2 - 1 - 2 361 103
          AF184073.1 - 1 - 2 349 103
          AF310103.1 - 1 - 2 275 103
          AF384665.1 - 1 - 2 328 103
          AK000211.1 - 1 - 2 313 103
          AK314422.1 - 1 - 2 353 103
          BC013569.1 - 1 - 2 268 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600010011.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16001052.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protease Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PYCARD

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          myeloid dendritic cell activation
          activation of innate immune response
          myeloid dendritic cell activation involved in immune response
          positive regulation of antigen processing and presentation of peptide antigen via MHC class II
          positive regulation of adaptive immune response
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          inflammatory response
          signal transduction
          regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of actin filament polymerization
          regulation of protein stability
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of interferon-beta production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          tumor necrosis factor-mediated signaling pathway
          positive regulation of activated T cell proliferation
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of apoptotic process
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          macropinocytosis
          innate immune response
          positive regulation of JNK cascade
          positive regulation of interleukin-1 beta secretion
          positive regulation of phagocytosis
          defense response to Gram-negative bacterium
          positive regulation of T cell activation
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of NF-kappaB transcription factor activity
          defense response to virus
          positive regulation of ERK1 and ERK2 cascade
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          negative regulation of protein serine/threonine kinase activity
          intrinsic apoptotic signaling pathway by p53 class mediator
          positive regulation of chemokine secretion
          positive regulation of release of cytochrome c from mitochondria
          positive regulation of T cell migration
          positive regulation of interleukin-8 secretion
          positive regulation of interleukin-6 secretion
          positive regulation of cysteine-type endopeptidase activity
          positive regulation of interleukin-10 secretion
          positive regulation of extrinsic apoptotic signaling pathway
          regulation of intrinsic apoptotic signaling pathway
          protease binding
          interleukin-6 receptor binding
          protein binding
          tropomyosin binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          myosin I binding
          enzyme binding
          Pyrin domain binding
          identical protein binding
          protein homodimerization activity
          BMP receptor binding
          cysteine-type endopeptidase activity involved in apoptotic process

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR16015027.hg.1 ENST00000247470.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015027.hg.1 ENST00000350605.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015027.hg.1 NM_013258.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015027.hg.1 NM_145182.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015027.hg.1 TC16001052.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1600007524.hg.1 ENST00000561916.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007524.hg.1 ENST00000561916.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007524.hg.1 ENST00000561916.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007524.hg.1 ENST00000561916.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007524.hg.1 NR_102400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007524.hg.1 NR_102400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007524.hg.1 NR_102400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007524.hg.1 NR_102400.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015029.hg.1 ENST00000247470.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015029.hg.1 ENST00000350605.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015029.hg.1 NM_013258.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015029.hg.1 NM_145182.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015029.hg.1 TC16001052.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000350605.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000350605.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000350605.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000350605.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000350605.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000565022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000565022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000565022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000565022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 ENST00000565022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 NM_013258.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 NM_013258.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 NM_013258.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 NM_013258.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 NM_013258.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 NM_013258.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 NM_145182.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 NM_145182.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 NM_145182.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 NM_145182.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600010011.hg.1 NM_145182.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015028.hg.1 ENST00000247470.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015028.hg.1 ENST00000350605.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015028.hg.1 ENST00000355986.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015028.hg.1 NM_013258.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015028.hg.1 NM_145182.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015028.hg.1 TC16001052.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR16015025.hg.1 ENST00000247470.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015025.hg.1 ENST00000355986.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16015025.hg.1 NM_013258.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16015025.hg.1 TC16001052.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000350605.4 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000350605.4 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000350605.4 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000350605.4 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000350605.4 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000565022.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000565022.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000565022.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000565022.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 ENST00000565022.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 NM_013258.4 Refseq
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 NM_013258.4 Refseq
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 NM_013258.4 Refseq
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 NM_013258.4 Refseq
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 NM_013258.4 Refseq
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 NM_013258.4 Refseq
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 NM_145182.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 NM_145182.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 NM_145182.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 NM_145182.2 Refseq
          Affymetrix Human Clariom D Assay TC1600010011.hg.1 NM_145182.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600177488.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1600177488.hg.1 ENST00000350605.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1600177488.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600177488.hg.1 NM_013258.4 Refseq
          Affymetrix Human Clariom D Assay PSR1600177488.hg.1 NM_145182.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600177488.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1600177488.hg.1 ENST00000350605.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1600177488.hg.1 ENST00000561508.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600177488.hg.1 NM_013258.4 Refseq
          Affymetrix Human Clariom D Assay PSR1600177488.hg.1 NM_145182.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600177488.hg.1 TC1600010011.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1600007524.hg.1 ENST00000561916.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600007524.hg.1 ENST00000561916.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600007524.hg.1 ENST00000561916.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600007524.hg.1 ENST00000561916.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600007524.hg.1 NR_102400.1 Refseq
          Affymetrix Human Clariom D Assay TC1600007524.hg.1 NR_102400.1 Refseq
          Affymetrix Human Clariom D Assay TC1600007524.hg.1 NR_102400.1 Refseq
          Affymetrix Human Clariom D Assay TC1600007524.hg.1 NR_102400.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600177482.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1600177482.hg.1 ENST00000565022.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600177482.hg.1 NM_013258.4 Refseq
          Affymetrix Human Clariom D Assay PSR1600177482.hg.1 ENST00000247470.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1600177482.hg.1 ENST00000565022.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600177482.hg.1 NM_013258.4 Refseq
          Affymetrix Human Clariom D Assay PSR1600177482.hg.1 TC1600010011.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline