| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000359634.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000409086.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000412905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000412905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000412905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000412905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000418005.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000418005.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000418005.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000430412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000430412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000430412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000430412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000430412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000430412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000430412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000430412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000430412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000430412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000430412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000430412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000430412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000430412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000444421.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000444421.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000444421.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000444421.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000458054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000458054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000458054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000458054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000464301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000464301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000465851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000465851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000465851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000465851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000465851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000465851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000465851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000465851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
ENST00000465851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_001127257.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010328.hg.1 |
NM_020342.2 |
Refseq |