Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01548831_m1
          See other TMED3 GE Assays ›
          Gene Symbol:
          TMED3
          Gene Name:
          transmembrane p24 trafficking protein 3
          Gene Aliases:
          C15orf22, P24B, p24g4, p24gamma4, p26
          Chromosome Location:
          Chr.15: 79311112 - 79413886 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01548831_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TMED3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001301203.3 NP_001288132.1 1 - 2 305 114
          NM_001330376.2 NP_001317305.1 1 - 2 305 114
          NM_001363735.2 NP_001350664.1 1 - 2 305 114
          NM_007364.4 NP_031390.1 1 - 2 305 114
          NR_125394.2 1 - 2 305 114
          GenBank mRNA AJ132270.1 - 1 - 2 186 114
          AK075529.1 - 1 - 2 258 114
          AK222516.1 - 1 - 2 293 114
          AK289434.1 - 1 - 2 274 114
          AK297710.1 - 1 - 2 272 114
          AK304148.1 - 1 - 2 261 114
          AL109672.1 - 1 - 2 265 114
          AY358974.1 - 1 - 2 265 114
          BC000027.2 - 1 - 2 263 114
          BC010853.1 - 1 - 2 300 114
          BC017495.1 - 1 - 2 266 114
          BC022232.1 - 1 - 2 290 114
          BC111547.1 - 1 - 2 326 114
          BM549126.1 - 1 - 2 192 114
          BP260160.1 - 1 - 2 355 114
          CR457384.1 - 1 - 2 167 114
          HM005671.1 - 1 - 2 268 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500008003.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15000741.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TMED3

          Gene Ontology Categories:

          Function(s)

          intracellular protein transport
          ER to Golgi vesicle-mediated transport
          Golgi organization
          protein transport

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR15006674.hg.1 ENST00000299705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15006674.hg.1 ENST00000424155.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15006674.hg.1 ENST00000536821.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15006674.hg.1 NM_007364.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15006674.hg.1 TC15000741.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15006675.hg.1 ENST00000299705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15006675.hg.1 ENST00000424155.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15006675.hg.1 ENST00000536821.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15006675.hg.1 NM_007364.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15006675.hg.1 TC15000741.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000558562.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000558562.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000558562.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000558562.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000561560.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_007364.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_007364.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_007364.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_007364.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_007364.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_007364.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_007364.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000558562.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000558562.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000558562.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000558562.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000561560.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_007364.3 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_007364.3 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_007364.3 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_007364.3 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_007364.3 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_007364.3 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_007364.3 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157557.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157557.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157557.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157557.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157557.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157557.hg.1 NM_007364.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500157557.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157557.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157557.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157557.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157557.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157557.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157557.hg.1 NM_007364.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500157557.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157557.hg.1 TC1500008003.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500157547.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157547.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157547.hg.1 NM_007364.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500157547.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157547.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157547.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157547.hg.1 NM_007364.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500157547.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157547.hg.1 TC1500008003.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500157556.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157556.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157556.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157556.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157556.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157556.hg.1 NM_007364.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500157556.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157556.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157556.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157556.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157556.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157556.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157556.hg.1 NM_007364.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500157556.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157556.hg.1 TC1500008003.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500157555.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157555.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157555.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157555.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157555.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157555.hg.1 NM_007364.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500157555.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157555.hg.1 ENST00000299705.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157555.hg.1 ENST00000424155.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157555.hg.1 ENST00000536821.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157555.hg.1 ENST00000543455.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500157555.hg.1 NM_001301203.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157555.hg.1 NM_007364.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500157555.hg.1 NR_125394.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500157555.hg.1 TC1500008003.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 215628 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline