| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016749.hg.1 |
ENST00000375543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016749.hg.1 |
ENST00000375544.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016749.hg.1 |
ENST00000395538.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016749.hg.1 |
NM_001193335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016749.hg.1 |
NM_017680.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016749.hg.1 |
TC09001336.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016752.hg.1 |
ENST00000375543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016752.hg.1 |
ENST00000375544.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016752.hg.1 |
ENST00000395538.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016752.hg.1 |
ENST00000450139.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016752.hg.1 |
NM_001193335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016752.hg.1 |
NM_017680.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016752.hg.1 |
TC09001336.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016759.hg.1 |
NM_001193335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016759.hg.1 |
NM_017680.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016759.hg.1 |
TC09001336.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016756.hg.1 |
ENST00000375543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016756.hg.1 |
ENST00000375544.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016756.hg.1 |
ENST00000395538.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016756.hg.1 |
ENST00000450139.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016756.hg.1 |
NM_001193335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016756.hg.1 |
NM_017680.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016756.hg.1 |
TC09001336.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016755.hg.1 |
ENST00000375543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016755.hg.1 |
ENST00000375544.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016755.hg.1 |
ENST00000395538.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016755.hg.1 |
ENST00000450139.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016755.hg.1 |
NM_001193335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016755.hg.1 |
NM_017680.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016755.hg.1 |
TC09001336.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016758.hg.1 |
ENST00000375543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016758.hg.1 |
ENST00000375544.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016758.hg.1 |
ENST00000395538.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016758.hg.1 |
ENST00000450139.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016758.hg.1 |
NM_001193335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016758.hg.1 |
NM_017680.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016758.hg.1 |
TC09001336.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016753.hg.1 |
ENST00000375543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016753.hg.1 |
ENST00000375544.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016753.hg.1 |
ENST00000395538.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016753.hg.1 |
ENST00000450139.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016753.hg.1 |
NM_001193335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016753.hg.1 |
NM_017680.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016753.hg.1 |
TC09001336.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900173520.hg.1 |
NM_001193335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173520.hg.1 |
NM_017680.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173520.hg.1 |
NM_001193335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173520.hg.1 |
NM_017680.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173520.hg.1 |
TC0900010797.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900173517.hg.1 |
ENST00000375543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173517.hg.1 |
ENST00000375544.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173517.hg.1 |
NM_001193335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173517.hg.1 |
NM_017680.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173517.hg.1 |
ENST00000375543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173517.hg.1 |
ENST00000375544.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173517.hg.1 |
NM_001193335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173517.hg.1 |
NM_017680.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173517.hg.1 |
TC0900010797.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900173518.hg.1 |
ENST00000375543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173518.hg.1 |
ENST00000375544.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173518.hg.1 |
NM_001193335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173518.hg.1 |
NM_017680.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173518.hg.1 |
ENST00000375543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173518.hg.1 |
ENST00000375544.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173518.hg.1 |
NM_001193335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173518.hg.1 |
NM_017680.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173518.hg.1 |
TC0900010797.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900173519.hg.1 |
ENST00000375543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173519.hg.1 |
ENST00000375544.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173519.hg.1 |
NM_001193335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173519.hg.1 |
NM_017680.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173519.hg.1 |
ENST00000375543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173519.hg.1 |
ENST00000375544.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173519.hg.1 |
NM_001193335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173519.hg.1 |
NM_017680.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173519.hg.1 |
TC0900010797.hg.1 |
Clariom D TC |