| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000479125.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000479125.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000479125.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000479125.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000479125.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000487953.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000487953.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000487953.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000487953.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000487953.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000490098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000490098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000490098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000490098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000490098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
ENST00000479125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
ENST00000487953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
ENST00000490098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
ENST00000479125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
ENST00000487953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
ENST00000490098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188518.hg.1 |
TC0100011999.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000479125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000479125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000479125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000479125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000479125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000487953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000487953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000487953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000487953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000487953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000490098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000490098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000490098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000490098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000490098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
ENST00000496662.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011999.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188516.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188516.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188516.hg.1 |
ENST00000479125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188516.hg.1 |
ENST00000487953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188516.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188516.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188516.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188516.hg.1 |
ENST00000366627.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188516.hg.1 |
ENST00000366628.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188516.hg.1 |
ENST00000479125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188516.hg.1 |
ENST00000487953.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188516.hg.1 |
ENST00000490807.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188516.hg.1 |
ENST00000494689.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100188516.hg.1 |
NM_032324.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100188516.hg.1 |
TC0100011999.hg.1 |
Clariom D TC |