Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01552756_m1
          See other BPI GE Assays ›
          Gene Symbol:
          BPI
          Gene Name:
          bactericidal permeability increasing protein
          Gene Aliases:
          BPIFD1, rBPI
          Chromosome Location:
          Chr.20: 38304156 - 38337505 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01552756_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BPI
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001725.3 NP_001716.3 9 - 10 1063 96
          XM_024451972.2 XP_024307740.1 5 - 6 489 96
          XM_047440393.1 XP_047296349.1 9 - 10 1063 96
          XM_047440394.1 XP_047296350.1 9 - 10 1063 96
          XM_047440395.1 XP_047296351.1 5 - 6 489 96
          GenBank mRNA AF322588.1 - 9 - 10 995 96
          AK296568.1 - 9 - 10 914 96
          AK315328.1 - 9 - 10 1036 96
          AX713278.1 - 9 - 10 1037 96
          AX746123.1 - 9 - 10 1037 96
          AX752831.1 - 9 - 10 1037 96
          BC040955.1 - 9 - 10 1044 96
          DQ414688.1 - 9 - 10 1007 96
          J04739.1 - 9 - 10 1037 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000009918.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000281.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BPI

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          antibacterial humoral response
          lipopolysaccharide-mediated signaling pathway
          negative regulation of interleukin-6 production
          negative regulation of interleukin-8 production
          negative regulation of tumor necrosis factor production
          defense response to bacterium
          negative regulation of macrophage activation
          innate immune response
          defense response to Gram-negative bacterium
          antimicrobial humoral immune response mediated by antimicrobial peptide
          cellular response to lipopolysaccharide
          lipopolysaccharide binding
          lipid binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000489039.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000489039.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000489039.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000489039.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000490381.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000490381.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000490381.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000490381.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 ENST00000490381.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20003999.hg.1 ENST00000262865.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20003999.hg.1 ENST00000489102.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20003999.hg.1 NM_001725.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20003999.hg.1 TC20000281.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000489039.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000489039.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000489039.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000489039.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000490381.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000490381.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000490381.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000490381.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 ENST00000490381.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay TC2000009918.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000169778.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2000169778.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2000169778.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000169778.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000169778.hg.1 ENST00000262865.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2000169778.hg.1 ENST00000417318.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2000169778.hg.1 ENST00000489102.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000169778.hg.1 NM_001725.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000169778.hg.1 TC2000009918.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 145623 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline