Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01552822_m1
          See other NTNG1 GE Assays ›
          Gene Symbol:
          NTNG1
          Gene Name:
          netrin G1
          Gene Aliases:
          Lmnt1, NetG1, NetrinG1
          Chromosome Location:
          Chr.1: 107140088 - 107484923 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01552822_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NTNG1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113226.3 NP_001106697.1 2 - 3 878 61
          NM_001113228.3 NP_001106699.1 2 - 3 901 61
          NM_001312688.2 NP_001299617.1 2 - 3 888 61
          NM_001330665.2 NP_001317594.1 2 - 3 878 61
          NM_001372166.1 NP_001359095.1 2 - 3 878 61
          NM_001372167.1 NP_001359096.1 2 - 3 888 61
          NM_001372168.1 NP_001359097.1 2 - 3 878 61
          NM_001372169.1 NP_001359098.1 2 - 3 878 61
          NM_001372170.1 NP_001359099.1 2 - 3 901 61
          NM_001372171.1 NP_001359100.1 2 - 3 901 61
          NM_014917.4 NP_055732.2 2 - 3 888 61
          XM_011541025.3 XP_011539327.1 2 - 3 878 61
          XM_017000683.3 XP_016856172.1 2 - 3 888 61
          XM_017000686.3 XP_016856175.1 2 - 3 888 61
          XM_047449446.1 XP_047305402.1 2 - 3 1399 61
          XM_047449447.1 XP_047305403.1 2 - 3 888 61
          XM_047449448.1 XP_047305404.1 2 - 3 901 61
          XM_047449449.1 XP_047305405.1 2 - 3 888 61
          GenBank mRNA AB023193.1 - 2 - 3 990 61
          AK296533.1 - 2 - 3 959 61
          AY358365.1 - 1 - 2 516 61
          BC030220.1 - 2 - 3 853 61
          BX538348.1 - 2 - 3 969 61
          KJ534901.1 - 1 - 2 251 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100009307.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000926.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NTNG1

          Gene Ontology Categories:

          Function(s) Process(es)

          nervous system development
          axonogenesis
          regulation of neuron projection development
          cell differentiation
          modulation of synaptic transmission
          synaptic membrane adhesion
          regulation of neuron projection arborization
          regulation of neuron migration
          protein binding
          cell adhesion molecule binding
          protein binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000294649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000294649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000294649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000294649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000294649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000294649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000294649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000294649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000294649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000294649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000294649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370066.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370066.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370066.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370066.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370066.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370066.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370066.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370066.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370066.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370066.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370067.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370068.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370071.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370073.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370073.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370073.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370073.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370073.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370073.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370073.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370073.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370073.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370073.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370073.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370073.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370073.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370073.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370073.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370074.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370074.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370074.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370074.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370074.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370074.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370074.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370074.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370074.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370074.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370074.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370074.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370074.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370076.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000370076.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000462149.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000462149.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000462149.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000462149.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000477948.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000477948.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000477948.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000477948.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 ENST00000477948.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_001113228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_014917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_014917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_014917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_014917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_014917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_014917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_014917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_014917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_014917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_014917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_014917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009307.hg.1 NM_014917.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 200177 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline