Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301668.1 | NP_001288597.1 | 7 - 8 | 1295 | 79 | |
NM_015878.5 | NP_056962.2 | 8 - 9 | 1441 | 79 | ||
NM_148174.3 | NP_680479.1 | 7 - 8 | 1295 | 79 | ||
XM_005250969.1 | XP_005251026.1 | 7 - 8 | 1256 | 79 | ||
XM_011517126.1 | XP_011515428.1 | 8 - 9 | 1428 | 79 | ||
XM_011517127.1 | XP_011515429.1 | 8 - 9 | 1419 | 79 | ||
XM_011517128.2 | XP_011515430.1 | 7 - 8 | 1439 | 79 | ||
XM_017013563.1 | XP_016869052.1 | 8 - 9 | 1407 | 79 | ||
XM_017013564.1 | XP_016869053.1 | 8 - 9 | 1419 | 79 | ||
XM_017013565.1 | XP_016869054.1 | 8 - 9 | 1558 | 79 | ||
XM_017013566.1 | XP_016869055.1 | 7 - 8 | 1439 | 79 | ||
GenBank mRNA | AK095493.1 | - | 7 - 8 | 1223 | 79 | |
AK290516.1 | - | 8 - 9 | 1173 | 79 | ||
BC013420.2 | - | 8 - 9 | 1380 | 3509827 | 79 | |
BC019279.1 | - | 7 - 8 | 1242 | 2823000 | 79 | |
BC042404.1 | - | 7 - 8 | 1266 | 79 | ||
BT006858.1 | - | 5 - 6 | 668 | 79 | ||
CR456745.1 | - | 5 - 6 | 668 | 79 | ||
D88674.1 | - | 8 - 9 | 1377 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |