Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01558200_m1
          See other NOX4 GE Assays ›
          Gene Symbol:
          NOX4 CDKN1B
          Gene Name:
          NADPH oxidase 4
          cyclin dependent kinase inhibitor 1B
          Gene Aliases:
          KOX, KOX-1, RENOX
          CDKN4, KIP1, MEN1B, MEN4, P27KIP1
          Chromosome Location:
          Chr.11: 89324353 - 89589557 on Build GRCh38
          Chromosome Location:
          Chr.12: 12717368 - 12722369 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs01558200_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NOX4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001143836.3 NP_001137308.2 15 - 16 1522 93
          NM_001143837.2 NP_001137309.2 19 - 20 1923 93
          NM_001291926.2 NP_001278855.2 16 - 17 1638 93
          NM_001291927.1 NP_001278856.1 16 - 17 1582 93
          NM_001291929.2 NP_001278858.2 15 - 16 1514 93
          NM_001300995.1 NP_001287924.1 15 - 16 1435 93
          NM_016931.5 NP_058627.2 16 - 17 1642 93
          GenBank mRNA AB041035.1 - 16 - 17 1752 93
          AF254621.1 - 16 - 17 1599 93
          AF261943.1 - 16 - 17 1513 93
          AJ704725.1 - 16 - 17 1509 93
          AJ704726.1 - 15 - 16 1393 93
          AJ704727.1 - 13 - 14 1068 93
          AJ704728.1 - 7 - 8 592 93
          AJ704729.1 - 6 - 7 472 93
          AK057189.1 - 19 - 20 1977 93
          AK291830.1 - 15 - 16 1475 93
          AK298323.1 - 15 - 16 1406 93
          AK298328.1 - 16 - 17 1582 93
          AK298357.1 - 15 - 16 1486 93
          AK298376.1 - 12 - 13 1210 93
          AY288918.1 - 15 - 16 1393 93
          BC040105.1 - 16 - 17 1585 93
          Gene Symbol: CDKN1B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AJ616234.1 - 6 - 7 472 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100011901.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11002178.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NOX4
          Click here to find siRNAs for Gene CDKN1B

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          heart process
          superoxide metabolic process
          defense response
          inflammatory response
          negative regulation of cell proliferation
          gene expression
          intracellular signal transduction
          superoxide anion generation
          bone resorption
          homocysteine metabolic process
          positive regulation of protein kinase B signaling
          cardiac muscle cell differentiation
          positive regulation of ERK1 and ERK2 cascade
          cellular response to glucose stimulus
          reactive oxygen species metabolic process
          reactive oxygen species biosynthetic process
          positive regulation of DNA biosynthetic process
          nucleotide binding
          protein binding
          electron carrier activity
          NAD(P)H oxidase activity
          superoxide-generating NADPH oxidase activity
          oxidoreductase activity
          oxygen sensor activity
          heme binding
          flavin adenine dinucleotide binding
          modified amino acid binding
          superoxide-generating NADPH oxidase activity
          protein tyrosine kinase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 ENST00000263317.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 ENST00000343727.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 ENST00000375979.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 ENST00000413594.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 ENST00000424319.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 ENST00000525196.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 ENST00000527626.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 ENST00000527956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 ENST00000528341.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 ENST00000531342.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 ENST00000532825.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 ENST00000534731.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 ENST00000535633.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 ENST00000542487.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 NM_001143836.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 NM_001143837.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 NM_016931.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11026082.hg.1 TC11002178.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000263317.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000343727.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000375979.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000424319.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000524473.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000525196.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000527626.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000527956.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000528341.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000529343.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000531342.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000532825.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000534731.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 NM_001143836.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 NM_001143837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 NM_001291926.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 NM_001291927.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 NM_001291929.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 NM_001300995.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 NM_016931.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000263317.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000343727.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000375979.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000424319.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000524473.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000525196.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000527626.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000527956.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000528341.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000529343.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000531342.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000532825.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 ENST00000534731.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 NM_001143836.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 NM_001143837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 NM_001291926.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 NM_001291927.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 NM_001291929.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 NM_001300995.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 NM_016931.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100191883.hg.1 TC1100011901.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0