| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05002291.hg.1 |
TC05000193.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05002291.hg.1 |
ENST00000361970.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05002291.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500149134.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149134.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500149134.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149134.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500149134.hg.1 |
TC0500007267.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000361970.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
ENST00000388827.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007267.hg.1 |
NM_001134848.1 |
Refseq |