| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08010807.hg.1 |
ENST00000330824.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08010807.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08010807.hg.1 |
TC08000816.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
NM_001301772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
NM_001301772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
NM_001301772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
NM_001301772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
NM_001301772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
NM_001301772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800162481.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800162481.hg.1 |
NM_001301772.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800162481.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800162481.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800162481.hg.1 |
NM_001301772.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800162481.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800162481.hg.1 |
TC0800009186.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
ENST00000622500.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
NM_001301772.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
NM_001301772.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
NM_001301772.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
NM_001301772.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
NM_001301772.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
NM_001301772.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009186.hg.1 |
NM_178172.5 |
Refseq |