| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000546639.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000546639.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000546639.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000546639.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000546785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000546785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000546785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000547342.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000547342.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000547342.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000547342.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000547342.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000547342.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000552280.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000552280.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000552280.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12026136.hg.1 |
ENST00000257549.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12026136.hg.1 |
ENST00000446302.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12026136.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12026136.hg.1 |
TC12001989.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200193434.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200193434.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200193434.hg.1 |
TC1200011994.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200193436.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200193436.hg.1 |
ENST00000552280.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200193436.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200193436.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200193436.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200193436.hg.1 |
ENST00000552280.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200193436.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200193436.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200193436.hg.1 |
TC1200011994.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200193432.hg.1 |
TC1200011994.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000257549.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000546639.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000546639.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000546639.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000546639.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000546785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000546785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000546785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000547342.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000547342.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000547342.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000547342.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000547342.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000547342.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000552280.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000552280.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000552280.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
ENST00000553112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011994.hg.1 |
NM_006843.2 |
Refseq |