Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01569163_m1
          See other IMMP2L GE Assays ›
          Gene Symbol:
          IMMP2L
          Gene Name:
          inner mitochondrial membrane peptidase subunit 2
          Gene Aliases:
          IMMP2L-IT1, IMP2, IMP2-LIKE
          Chromosome Location:
          Chr.7: 110662645 - 111562531 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01569163_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IMMP2L
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001244606.1 NP_001231535.1 3 - 4 264 83
          NM_032549.3 NP_115938.1 2 - 3 578 83
          XM_005250630.3 XP_005250687.1 4 - 5 346 83
          XM_011516605.2 XP_011514907.1 3 - 4 278 83
          XM_011516608.2 XP_011514910.1 2 - 3 540 83
          XM_011516609.2 XP_011514911.1 2 - 3 540 83
          XM_011516611.2 XP_011514913.1 2 - 3 540 83
          XM_011516613.2 XP_011514915.1 2 - 3 540 83
          XM_017012699.1 XP_016868188.1 2 - 3 540 83
          XM_017012700.1 XP_016868189.1 3 - 4 278 83
          XM_017012701.1 XP_016868190.1 3 - 4 229 83
          XM_017012702.1 XP_016868191.1 4 - 5 346 83
          XM_017012703.1 XP_016868192.1 2 - 3 540 83
          XM_017012704.1 XP_016868193.1 2 - 3 540 83
          XM_017012705.1 XP_016868194.1 3 - 4 316 83
          GenBank mRNA AF359563.1 - 2 - 3 578 83
          AK311968.1 - 1 - 2 229 83
          BC008497.1 - 3 - 4 229 4279910 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700012255.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001758.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IMMP2L

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          signal peptide processing
          protein processing involved in protein targeting to mitochondrion
          superoxide metabolic process
          cellular response to DNA damage stimulus
          spermatogenesis
          brain development
          blood circulation
          respiratory electron transport chain
          ovulation
          mitochondrial respiratory chain complex assembly
          cerebellum vasculature development
          serine-type endopeptidase activity
          peptidase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07027476.hg.1 TC07001758.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07027476.hg.1 ENST00000331762.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07027476.hg.1 ENST00000405709.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07027476.hg.1 ENST00000415362.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07027476.hg.1 ENST00000437687.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07027476.hg.1 ENST00000447215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07027476.hg.1 ENST00000452895.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07027476.hg.1 NM_001244606.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07027476.hg.1 NM_032549.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 ENST00000331762.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 ENST00000405709.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 ENST00000437687.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 ENST00000447215.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 ENST00000452753.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 ENST00000452895.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 ENST00000492938.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 NM_001244606.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 NM_032549.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 ENST00000331762.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 ENST00000405709.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 ENST00000437687.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 ENST00000447215.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 ENST00000452753.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 ENST00000452895.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 ENST00000492938.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 NM_001244606.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 NM_032549.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700188455.hg.1 TC0700012255.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline