| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004218.hg.1 |
ENST00000314529.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004218.hg.1 |
ENST00000418905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004218.hg.1 |
NM_001256167.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004218.hg.1 |
NM_001256168.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004218.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004218.hg.1 |
TC03000211.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000418905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000418905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000418905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000418905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000418905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000433723.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000433723.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000433723.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
ENST00000433723.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_001256167.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_001256167.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_001256167.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_001256167.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_001256167.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_001256167.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_001256168.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_001256168.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_001256168.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_001256168.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_001256168.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_001256168.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009612.hg.1 |
ENST00000558749.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009612.hg.1 |
ENST00000558749.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004217.hg.1 |
ENST00000314529.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004217.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03004217.hg.1 |
TC03000211.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000418905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000418905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000418905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000418905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000418905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000433723.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000433723.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000433723.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
ENST00000433723.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_001256167.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_001256167.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_001256167.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_001256167.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_001256167.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_001256167.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_001256168.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_001256168.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_001256168.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_001256168.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_001256168.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_001256168.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007123.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009612.hg.1 |
ENST00000558749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009612.hg.1 |
ENST00000558749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300150103.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300150103.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300150103.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300150103.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300150103.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300150103.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300150103.hg.1 |
TC0300007123.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300150104.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300150104.hg.1 |
ENST00000418905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300150104.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300150104.hg.1 |
NM_001256167.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300150104.hg.1 |
NM_001256168.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300150104.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300150104.hg.1 |
ENST00000314529.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300150104.hg.1 |
ENST00000418905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300150104.hg.1 |
ENST00000420891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300150104.hg.1 |
NM_001256167.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300150104.hg.1 |
NM_001256168.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300150104.hg.1 |
NM_175888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300150104.hg.1 |
TC0300007123.hg.1 |
Clariom D TC |