| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027159.hg.1 |
ENST00000335230.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027159.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027159.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027159.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027159.hg.1 |
TC05001950.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027160.hg.1 |
ENST00000335230.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027160.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027160.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027160.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027160.hg.1 |
TC05001950.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012504.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187268.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187268.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187268.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187268.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187268.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187268.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187268.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187268.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187268.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187268.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187268.hg.1 |
TC0500012504.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012508.hg.1 |
ENST00000517788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187269.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187269.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187269.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187269.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187269.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187269.hg.1 |
ENST00000335230.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187269.hg.1 |
ENST00000377713.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187269.hg.1 |
ENST00000423071.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187269.hg.1 |
NM_001145017.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187269.hg.1 |
NM_181774.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187269.hg.1 |
TC0500012504.hg.1 |
Clariom D TC |