| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000551896.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000551896.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000551896.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000551896.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000551896.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
ENST00000551896.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12005538.hg.1 |
ENST00000301466.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12005538.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12005538.hg.1 |
TC12000429.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000551896.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000551896.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000551896.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000551896.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000551896.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
ENST00000551896.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007694.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200154992.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200154992.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200154992.hg.1 |
ENST00000551896.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200154992.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200154992.hg.1 |
ENST00000301466.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200154992.hg.1 |
ENST00000542365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200154992.hg.1 |
ENST00000551896.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200154992.hg.1 |
NM_003578.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200154992.hg.1 |
TC1200007694.hg.1 |
Clariom D TC |