Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01574154_m1
          See other IL22 GE Assays ›
          Gene Symbol:
          IL22
          Gene Name:
          interleukin 22
          Gene Aliases:
          IL-21, IL-22, IL-D110, IL-TIF, ILTIF, TIFIL-23, TIFa, zcyto18
          Chromosome Location:
          Chr.12: 68248245 - 68253501 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01574154_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IL22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020525.4 NP_065386.1 3 - 4 445 66
          GenBank mRNA AF279437.1 - 3 - 4 449 66
          AJ277247.1 - 4 - 5 463 66
          AY358890.1 - 3 - 4 449 66
          BC066263.1 - 3 - 4 442 6971854 66
          BC066264.1 - 3 - 4 441 6971855 66
          BC066265.1 - 3 - 4 442 6971857 66
          BC067509.1 - 3 - 4 442 6971858 66
          BC067510.1 - 3 - 4 442 6971859 66
          BC069112.1 - 4 - 5 463 7216929 66
          BC069308.1 - 3 - 4 442 7262127 66
          BC070261.1 - 4 - 5 460 30416027 66
          CD559617.2 - 3 - 4 472 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200011179.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001698.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IL22

          Gene Ontology Categories:

          Function(s) Process(es)

          acute-phase response
          inflammatory response
          immune response
          cell-cell signaling
          positive regulation of JAK-STAT cascade
          response to glucocorticoid
          cytokine activity
          protein binding
          interleukin-22 receptor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12022653.hg.1 ENST00000328087.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12022653.hg.1 ENST00000538666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12022653.hg.1 NM_020525.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12022653.hg.1 TC12001698.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Clariom D Assay TC1200011179.hg.1 NM_020525.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200186715.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200186715.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200186715.hg.1 NM_020525.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200186715.hg.1 ENST00000328087.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200186715.hg.1 ENST00000538666.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200186715.hg.1 NM_020525.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200186715.hg.1 TC1200011179.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 158140 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline