| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000416172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000416172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000416172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000416172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000416172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000416172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000558680.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000558680.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000558680.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000559364.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000559364.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000559364.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
ENST00000559364.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15004838.hg.1 |
ENST00000326648.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15004838.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15004838.hg.1 |
TC15000593.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1500153596.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500153596.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500153596.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500153596.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500153596.hg.1 |
TC1500007554.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000326648.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000416172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000416172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000416172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000416172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000416172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000416172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000558680.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000558680.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000558680.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000559364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000559364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000559364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
ENST00000559364.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007554.hg.1 |
NM_015042.1 |
Refseq |