Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01584837_s1
          See other IFIT2 GE Assays ›
          Gene Symbol:
          IFIT2
          Gene Name:
          interferon induced protein with tetratricopeptide repeats 2
          Gene Aliases:
          G10P2, GARG-39, IFI-54, IFI-54K, IFI54, IFIT-2, ISG-54 K, ISG-54K, ISG54, P54, cig42
          Chromosome Location:
          Chr.10: 89301949 - 89309276 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01584837_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IFIT2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001547.4 NP_001538.4 2 2631 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000008396.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IFIT2

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic mitochondrial changes
          response to virus
          negative regulation of protein binding
          cellular response to interferon-alpha
          positive regulation of apoptotic process
          defense response to virus
          type I interferon signaling pathway
          RNA binding
          protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000008396.hg.1 ENST00000371826.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008396.hg.1 ENST00000371826.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008396.hg.1 ENST00000371826.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008396.hg.1 ENST00000371826.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008396.hg.1 ENST00000371826.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008396.hg.1 ENST00000611722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008396.hg.1 ENST00000611722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008396.hg.1 ENST00000611722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008396.hg.1 NM_001547.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008396.hg.1 NM_001547.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008396.hg.1 NM_001547.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008396.hg.1 NM_001547.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008396.hg.1 NM_001547.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008396.hg.1 ENST00000371826.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008396.hg.1 ENST00000371826.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008396.hg.1 ENST00000371826.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008396.hg.1 ENST00000371826.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008396.hg.1 ENST00000371826.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008396.hg.1 ENST00000611722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008396.hg.1 ENST00000611722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008396.hg.1 ENST00000611722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008396.hg.1 NM_001547.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008396.hg.1 NM_001547.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008396.hg.1 NM_001547.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008396.hg.1 NM_001547.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008396.hg.1 NM_001547.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000156919.hg.1 ENST00000371826.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156919.hg.1 ENST00000611722.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156919.hg.1 NM_001547.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000156919.hg.1 ENST00000371826.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156919.hg.1 ENST00000611722.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156919.hg.1 NM_001547.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000156919.hg.1 TC1000008396.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline