Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01586570_sH
          See other SLC35G3 GE Assays ›
          Gene Symbol:
          SLC35G3
          Gene Name:
          solute carrier family 35 member G3
          Gene Aliases:
          AMAC1, TMEM21A
          Chromosome Location:
          Chr.17: 35192520 - 35194393 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01586570_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC35G3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152462.2 NP_689675.1 1 400 148
          GenBank mRNA AK097473.1 - 1 365 148
          BC136422.1 - 1 400 148
          BC140700.1 - 1 400 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700010417.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001367.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC35G3

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30866444 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10299676 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30866447 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10299678 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30866452 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30866470 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10299669 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10299673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16972415 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10299670 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 166755 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 28326 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 238460 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 278971 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P389532 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P389532 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 43511 - -
          Agilent Human Genome CGH Microarray 44B A_14_P129515 - -
          Agilent Human Genome CGH Microarray 44A A_14_P129515 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 43631 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P59988 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P389532 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P389532 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30866470 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30866451 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16972415 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10299678 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30866452 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30866470 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16972415 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30866452 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30866451 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10299678 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P389532 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P129515 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10299680 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P10299678 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P129515 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P129515 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P389532 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 125029 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 702 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P129515 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10299678 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10299670 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10299676 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10299680 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P12633073 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P129515 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_152462_at - -
          Affymetrix Human Gene 1.0 ST Array 8014232 - -
          Affymetrix Human Gene 1.0 ST Array 8004429 - -
          Affymetrix Human Gene 1.0 ST Array 8014231 - -
          Affymetrix Human Gene 1.0 ST Array 8004430 - -
          Affymetrix Human Gene 1.0 ST Array 8020163 - -
          Affymetrix Human Gene 1.1 ST Array 8014230 - -
          Affymetrix Human Gene 1.1 ST Array 8020162 - -
          Affymetrix Human Gene 1.1 ST Array 8004428 - -
          Affymetrix HT HG-U133+ PM Array Plate 1553471_PM_at - -
          Affymetrix Human Genome U219 Array 11738314_PM_at - -
          Affymetrix Human Genome U219 Array 11738314_at - -
          Affymetrix Human Genome U219 Array 11738315_x_at - -
          Affymetrix Human Genome U219 Array 11738315_PM_x_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1700006750.hg.1 ENST00000412468.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006750.hg.1 ENST00000412468.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006750.hg.1 ENST00000412468.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006750.hg.1 ENST00000412468.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006750.hg.1 ENST00000412468.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700006750.hg.1 NM_001102614.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006750.hg.1 NM_001102614.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700006750.hg.1 NM_001102614.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010417.hg.1 ENST00000297307.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010417.hg.1 ENST00000297307.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010417.hg.1 ENST00000297307.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010417.hg.1 ENST00000297307.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010417.hg.1 NM_152462.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010417.hg.1 NM_152462.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010417.hg.1 NM_152462.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17018112.hg.1 ENST00000297307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17018112.hg.1 NM_152462.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17018112.hg.1 TC17001367.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0800006739.hg.1 ENST00000382435.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006739.hg.1 ENST00000382435.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006739.hg.1 ENST00000382435.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006739.hg.1 ENST00000382435.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006739.hg.1 ENST00000382435.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800006739.hg.1 NM_054028.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800006739.hg.1 NM_054028.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800006739.hg.1 NM_054028.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800006739.hg.1 NM_054028.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010417.hg.1 ENST00000297307.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700010417.hg.1 ENST00000297307.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700010417.hg.1 ENST00000297307.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700010417.hg.1 ENST00000297307.6 Ensembl
          Affymetrix Human Clariom D Assay TC1700010417.hg.1 NM_152462.2 Refseq
          Affymetrix Human Clariom D Assay TC1700010417.hg.1 NM_152462.2 Refseq
          Affymetrix Human Clariom D Assay TC1700010417.hg.1 NM_152462.2 Refseq
          Affymetrix Human Clariom D Assay TC1700006750.hg.1 ENST00000412468.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700006750.hg.1 ENST00000412468.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700006750.hg.1 ENST00000412468.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700006750.hg.1 ENST00000412468.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700006750.hg.1 ENST00000412468.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700006750.hg.1 NM_001102614.1 Refseq
          Affymetrix Human Clariom D Assay TC1700006750.hg.1 NM_001102614.1 Refseq
          Affymetrix Human Clariom D Assay TC1700006750.hg.1 NM_001102614.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700181700.hg.1 ENST00000297307.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700181700.hg.1 NM_152462.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700181700.hg.1 ENST00000297307.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700181700.hg.1 NM_152462.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700181700.hg.1 TC1700010417.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700181701.hg.1 ENST00000297307.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700181701.hg.1 NM_152462.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700181701.hg.1 ENST00000297307.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700181701.hg.1 NM_152462.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700181701.hg.1 TC1700010417.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0800006739.hg.1 ENST00000382435.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800006739.hg.1 ENST00000382435.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800006739.hg.1 ENST00000382435.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800006739.hg.1 ENST00000382435.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800006739.hg.1 ENST00000382435.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800006739.hg.1 NM_054028.1 Refseq
          Affymetrix Human Clariom D Assay TC0800006739.hg.1 NM_054028.1 Refseq
          Affymetrix Human Clariom D Assay TC0800006739.hg.1 NM_054028.1 Refseq
          Affymetrix Human Clariom D Assay TC0800006739.hg.1 NM_054028.1 Refseq
          Human Genome U133 Plus 2.0 Array 1553340_s_at - -
          Human Genome U133 Plus 2.0 Array 1553471_PM_at - -
          Human Genome U133 Plus 2.0 Array 1553471_at - -
          Human Genome U133 Plus 2.0 Array 1553340_PM_s_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_152462_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553471_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553471_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553471_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553471_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553471_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553471_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553471_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553471_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553471_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553471_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553471_at1 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008183 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008183 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1797432 NM_152462.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1797432 AK097473.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1797432 NM_152462.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1797432 AK097473.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1797432 NM_152462.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1797432 AK097473.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1797432 NM_152462.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1797432 AK097473.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1797432 NM_152462.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1797432 AK097473.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_22748974-S NM_152462.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_22748974-S AK097473.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_3576-S - -
          Illumina human-6 v1.0 expression beadchip 0006520270 - -
          Illumina humanRef-8 v1.0 expression beadchip 0006520270 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1797432 NM_152462.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1797432 AK097473.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1797432 NM_152462.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1797432 AK097473.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 1340672 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_3576 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1797432 NM_152462.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1797432 AK097473.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1797432 NM_152462.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1797432 AK097473.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1797432 NM_152462.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1797432 AK097473.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1797432 NM_152462.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1797432 AK097473.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1797432 NM_152462.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1797432 AK097473.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline