Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01587630_g1
          See other SLC25A38 GE Assays ›
          Gene Symbol:
          SLC25A38
          Gene Name:
          solute carrier family 25 member 38
          Gene Aliases:
          SIDBA2
          Chromosome Location:
          Chr.3: 39383370 - 39397351 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01587630_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC25A38
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001354798.2 NP_001341727.1 4 - 5 811 76
          NM_017875.4 NP_060345.2 4 - 5 811 76
          XM_011533869.3 XP_011532171.1 5 - 6 3846 76
          XM_024453611.2 XP_024309379.1 5 - 6 475 76
          XM_047448413.1 XP_047304369.1 5 - 6 4452 76
          XM_047448414.1 XP_047304370.1 5 - 6 4932 76
          XM_047448415.1 XP_047304371.1 5 - 6 4840 76
          XM_047448416.1 XP_047304372.1 5 - 6 1680 76
          GenBank mRNA AK000558.1 - 4 - 5 833 76
          AK225388.1 - 4 - 5 809 76
          BC013194.1 - 4 - 5 518 76
          CR457242.1 - 4 - 5 456 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300007100.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03000201.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC25A38

          Gene Ontology Categories:

          Function(s) Process(es)

          heme biosynthetic process
          erythrocyte differentiation
          glycine import into mitochondrion
          glycine transmembrane transporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03003954.hg.1 ENST00000273158.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03003954.hg.1 NM_017875.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03003954.hg.1 TC03000201.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03003955.hg.1 ENST00000273158.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03003955.hg.1 NM_017875.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03003955.hg.1 TC03000201.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay TC0300007100.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300149813.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149813.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149813.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300149813.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149813.hg.1 ENST00000431510.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149813.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300149813.hg.1 TC0300007100.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300149814.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149814.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300149814.hg.1 ENST00000273158.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0300149814.hg.1 NM_017875.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300149814.hg.1 TC0300007100.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline