| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000585522.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000585522.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000585522.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000585522.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000585522.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000586687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000586687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000586687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000586687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000586687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000586687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000587422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000587422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000587422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000587422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007821.hg.1 |
ENST00000605008.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004676.hg.1 |
ENST00000306346.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004676.hg.1 |
ENST00000399057.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004676.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004676.hg.1 |
TC18000454.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1800157684.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157684.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157684.hg.1 |
ENST00000586687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157684.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157684.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800157684.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157684.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157684.hg.1 |
ENST00000586687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157684.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157684.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800157684.hg.1 |
TC1800008459.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000306346.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000585522.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000585522.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000585522.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000585522.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000585522.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000586585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000586687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000586687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000586687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000586687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000586687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000586687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000587422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000587422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000587422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000587422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
ENST00000589332.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008459.hg.1 |
NM_145756.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007821.hg.1 |
ENST00000605008.1 |
Ensembl |